Assay Details
| Gene Symbol: AADACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026915.1 | 1 - 2 | 160 | 122 | ||
| GenBank mRNA | BC014344.1 | - | 1 - 2 | 160 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | AADACL2 antisense RNA 1 | AADACL2-AS1 | Anti-sense | Chr. 3 - 151751179 - 151928175 on NCBI Build GRCh38 | ||
| Downstream | arylacetamide deacetylase | AADAC |
CES5A1, DAC |
38.3kb | Sense | Chr. 3 - 151814008 - 151828488 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000828.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014808.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014808.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014808.hg.1 | ENST00000483636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014808.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014808.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014808.hg.1 | TC03000828.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014812.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014812.hg.1 | ENST00000483636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014812.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014812.hg.1 | TC03000828.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014807.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014807.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014807.hg.1 | ENST00000483636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014807.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014807.hg.1 | TC03000828.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165971.hg.1 | TC0300009190.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000496128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009190.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000463420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000471766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165970.hg.1 | TC0300009190.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | ENST00000462937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | ENST00000483636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | ENST00000561502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | NR_026915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165976.hg.1 | TC0300009190.hg.1 | Clariom D TC |

AADACP1
Genomic Map