This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00909957_m1
See other SLC7A7 GE Assays ›
Gene Symbol:
SLC7A7
Gene Name:
solute carrier family 7 member 7
Gene Aliases:
LAT3, LPI, MOP-2, Y+LAT1, y+LAT-1
Chromosome Location:
Chr.14: 22773222 - 22819811 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00909957_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011537299.1 XP_011535601.1 1 - 2 155 106
GenBank mRNA BX248291.1 - 1 - 2 90 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011537299.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of arginine metabolic process
protein complex assembly
cellular amino acid metabolic process
transport
amino acid transport
L-amino acid transport
leukocyte migration
L-alpha-amino acid transmembrane transport
basic amino acid transmembrane transport
basic amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t6srt:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0