This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00912281_m1
See other RBM45 GE Assays ›
Gene Symbol:
RBM45
Gene Name:
RNA binding motif protein 45
Gene Aliases:
DRB1, RB-1
Chromosome Location:
Chr.2: 178112409 - 178129656 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00912281_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM45
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152945.3 NP_694453.2 3 - 4 673 126
XM_005246287.4 XP_005246344.1 3 - 4 683 126
XM_017003320.1 XP_016858809.1 3 - 4 1116 126
XM_017003321.1 XP_016858810.1 3 - 4 1117 126
XM_017003322.1 XP_016858811.1 3 - 4 682 126
GenBank mRNA AB036991.1 - 3 - 4 595 126
AF526533.1 - 3 - 4 673 126
AK126575.1 - 3 - 4 531 126
BC066549.1 - 3 - 4 642 5748065 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010112.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001069.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM45

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
cell differentiation
nucleotide binding
RNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02015865.hg.1 TC02001069.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02015865.hg.1 ENST00000286070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02015865.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02015867.hg.1 TC02001069.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02015867.hg.1 ENST00000286070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02015867.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000464647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000464647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000464647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000424099.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000455903.6 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000464647.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000464647.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000464647.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay TC0200010112.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000493048.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay PSR0200170822.hg.1 TC0200010112.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 ENST00000286070.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 ENST00000424000.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 ENST00000616198.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 NM_152945.3 Refseq
Affymetrix Human Clariom D Assay PSR0200170819.hg.1 TC0200010112.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0