This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00912694_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs00912694_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX417242.2 - 2 Chr. 11 - 62576190 - 62577257 [-] on Build GRCh38 66
GenBankmRNA BX113801.1 - 1 54 Chr. 11 - 62576749 - 62577425 [+] on Build GRCh38 66
GenBankmRNA DB044655.1 - 1 Chr. 11 - 62573886 - 62576852 [-] on Build GRCh38 66
GenBankmRNA AK125351.1 - 1 2629 Chr. 11 - 62574174 - 62577586 [+] on Build GRCh38 66
GenBankmRNA AA010470.1 - 1 54 Chr. 11 - 62576754 - 62577218 [+] on Build GRCh38 66

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream terminal uridylyl transferase 1,
U6 snRNA-specific
TUT1 PAPD2,
RBM21,
STARPAP,
TUTase,
URLC6
Sense Chr. 11 - 62575045 - 62591637 on NCBI Build GRCh38
Downstream metastasis associated 1 family member 2 MTA2 MTA1L1,
PID
16.4kb Sense Chr. 11 - 62593203 - 62601840 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0