This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00913272_m1
See other ALDH18A1 GE Assays ›
Gene Symbol:
ALDH18A1
Gene Name:
aldehyde dehydrogenase 18 family member A1
Gene Aliases:
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
Chromosome Location:
Chr.10: 95605929 - 95656810 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00913272_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH18A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323413.1 NP_001310342.1 1 - 2 363 144
NM_001323415.1 NP_001310344.1 1 - 2 363 144
NM_001323416.1 NP_001310345.1 1 - 2 363 144
NM_001323418.1 NP_001310347.1 1 - 2 363 144

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001323413.1, NM_001323415.1, NM_001323416.1, NM_001323418.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glutamate metabolic process
proline biosynthetic process
ornithine biosynthetic process
cellular amino acid biosynthetic process
phosphorylation
citrulline biosynthetic process
oxidation-reduction process
L-proline biosynthetic process
glutamate 5-kinase activity
glutamate-5-semialdehyde dehydrogenase activity
protein binding
ATP binding
delta1-pyrroline-5-carboxylate synthetase activity
identical protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371221.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000371224.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000483788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000485428.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 ENST00000489386.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_001017423.1 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq
Affymetrix Human Clariom D Assay TC1000011499.hg.1 NM_002860.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0