This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00913522_m1
See other FAM65B GE Assays ›
Gene Symbol:
FAM65B
Gene Name:
family with sequence similarity 65 member B
Gene Aliases:
C6orf32, DFNB104, DIFF40, DIFF48, MYONAP, PL48
Chromosome Location:
Chr.6: 24804281 - 25042288 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00913522_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM65B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011515008.2 XP_011513310.1 2 - 3 275 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011515008.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
skeletal muscle fiber development
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline