This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916017_m1
See other NXPE4 GE Assays ›
Gene Symbol:
NXPE4
Gene Name:
neurexophilin and PC-esterase domain family member 4
Gene Aliases:
C11orf33, FAM55D
Chromosome Location:
Chr.11: 114570591 - 114596215 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916017_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017678.2 NP_060148.2 3 - 4 550 95
GenBank mRNA AY547307.1 - 1 - 2 119 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012382.hg
This assay targets the same location as HTA transcript cluster(s) TC11002314.hg

Additional Information:

This assay is in the 5'UTR region of the AY547307.1,NM_017678.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NXPE4

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11027676.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027676.hg.1 ENST00000424261.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027676.hg.1 NM_001077639.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11027676.hg.1 NM_017678.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11027676.hg.1 TC11002314.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay TC1100012382.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 ENST00000375478.3 Ensembl
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 ENST00000424261.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 NM_001077639.1 Refseq
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 NM_017678.2 Refseq
Affymetrix Human Clariom D Assay PSR1100195106.hg.1 TC1100012382.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0