This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916211_m1
See other HELB GE Assays ›
Gene Symbol:
HELB
Gene Name:
DNA helicase B
Gene Aliases:
DHB, hDHB
Chromosome Location:
Chr.12: 66302545 - 66343643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916211_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HELB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033647.4 NP_387467.2 5 - 6 1913 89
NR_135080.1 5 - 6 1913 89
XM_005269234.2 XP_005269291.1 5 - 6 1891 89
XM_017020230.1 XP_016875719.1 5 - 6 1891 89
GenBank mRNA AF319995.1 - 5 - 6 1854 89
AK290894.1 - 5 - 6 1878 89
BC143409.1 - 5 - 6 1879 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012655.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000593.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HELB

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
DNA replication
DNA replication, synthesis of RNA primer
DNA repair
cellular response to DNA damage stimulus
DNA duplex unwinding
regulation of DNA double-strand break processing
negative regulation of double-strand break repair via homologous recombination
ATP-dependent RNA helicase activity
ATP binding
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA helicase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000536862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000536862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000536862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0