This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916216_m1
See other HELB GE Assays ›
Gene Symbol:
HELB
Gene Name:
DNA helicase B
Gene Aliases:
DHB, hDHB
Chromosome Location:
Chr.12: 66302545 - 66343643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916216_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HELB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135081.1 4 - 5 1738 153
GenBank mRNA BC143411.1 - 4 - 5 1704 153
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK024464.1 - 1682 Chr. - - [-] on Build 153

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream interleukin 1 receptor associated kinase 3 IRAK3 ASRT5,
IRAKM
-55.1kb Sense Chr. 12 - 66189198 - 66254622 on NCBI Build GRCh38
Downstream glutamate receptor interacting protein 1 GRIP1 GRIP 32.0kb Anti-sense Chr. 12 - 66347431 - 67069265 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
DNA replication
DNA replication, synthesis of RNA primer
DNA repair
cellular response to DNA damage stimulus
DNA duplex unwinding
regulation of DNA double-strand break processing
negative regulation of double-strand break repair via homologous recombination
ATP-dependent RNA helicase activity
ATP binding
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA helicase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000440906.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000536862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000536862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000536862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000542394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 ENST00000545455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012655.hg.1 NM_033647.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0