This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916491_g1
See other POLR3D GE Assays ›
Gene Symbol:
POLR3D
Gene Name:
RNA polymerase III subunit D
Gene Aliases:
BN51T, RPC4, RPC53, TSBN51
Chromosome Location:
Chr.8: 22245104 - 22251167 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916491_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001722.2 NP_001713.2 4 - 5 442 112
GenBank mRNA AK026588.1 - 4 - 5 504 112
AY092086.1 - 3 - 4 359 112
BC002603.2 - 3 - 4 363 3143056 112
BC004484.2 - 3 - 4 385 3938075 112
CR536509.1 - 3 - 4 359 112
CR541803.1 - 3 - 4 359 112
M17754.1 - 4 - 5 401 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800006978.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000168.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR3D

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase III promoter
positive regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08001937.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001937.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001937.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08001945.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001945.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001945.hg.1 ENST00000397802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001945.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08001947.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001947.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001947.hg.1 ENST00000397802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001947.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08001949.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001949.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001949.hg.1 ENST00000397802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001949.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008077.hg.1 ENST00000398689.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08001948.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001948.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001948.hg.1 ENST00000397802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001948.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147493.hg.1 TC0800006978.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147496.hg.1 TC0800006978.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147495.hg.1 TC0800006978.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008077.hg.1 ENST00000398689.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147483.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147483.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147483.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147483.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147483.hg.1 TC0800006978.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147497.hg.1 TC0800006978.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0