This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916493_g1
See other POLR3D GE Assays ›
Gene Symbol:
POLR3D
Gene Name:
RNA polymerase III subunit D
Gene Aliases:
BN51T, RPC4, RPC53, TSBN51
Chromosome Location:
Chr.8: 22245104 - 22251167 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916493_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001722.2 NP_001713.2 6 - 7 735 72
GenBank mRNA AK026588.1 - 6 - 7 797 72
AY092086.1 - 5 - 6 652 72
BC002603.2 - 5 - 6 656 3143056 72
BC004484.2 - 5 - 6 678 3938075 72
CR536509.1 - 5 - 6 652 72
CR541803.1 - 5 - 6 652 72
M17754.1 - 6 - 7 694 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800006978.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000168.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR3D

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase III promoter
positive regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08001952.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001952.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001952.hg.1 ENST00000397802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001952.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08001951.hg.1 TC08000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08001951.hg.1 ENST00000306433.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001951.hg.1 ENST00000397802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08001951.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147499.hg.1 TC0800006978.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000518039.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 ENST00000519237.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay TC0800006978.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 ENST00000306433.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 ENST00000397802.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 ENST00000517789.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 NM_001722.2 Refseq
Affymetrix Human Clariom D Assay PSR0800147500.hg.1 TC0800006978.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0