Assay Details
| Gene Symbol: TM9SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004800.2 | NP_004791.1 | 3 - 4 | 560 | 105 | |
| GenBank mRNA | AK093507.1 | - | 3 - 4 | 259 | 105 | |
| AK290514.1 | - | 3 - 4 | 515 | 105 | ||
| AK315210.1 | - | 3 - 4 | 515 | 105 | ||
| BC110656.1 | - | 3 - 4 | 517 | 3855486 | 105 | |
| BC130293.1 | - | 3 - 4 | 394 | 40146324 | 105 | |
| U81006.1 | - | 3 - 4 | 460 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000356.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003753.hg.1 | ENST00000376387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003753.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003753.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003753.hg.1 | TC13000356.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152775.hg.1 | TC1300007843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000376387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000463709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | ENST00000466555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007843.hg.1 | NM_004800.2 | Refseq |

TM9SF2
Genomic Map