This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916785_m1
See other NDUFAF5 GE Assays ›
Gene Symbol:
NDUFAF5
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases:
C20orf7, bA526K24.2, dJ842G6.1
Chromosome Location:
Chr.20: 13785026 - 13821580 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916785_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFAF5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024120.4 NP_077025.2 5 - 6 512 104
NR_029377.1 5 - 6 512 104
XM_006723620.3 XP_006723683.1 5 - 6 489 104
XM_011529342.2 XP_011527644.1 5 - 6 489 104
GenBank mRNA AK091060.1 - 5 - 6 489 104
AK289781.1 - 5 - 6 504 104
BC005984.1 - 5 - 6 322 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC005984.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFAF5

Gene Ontology Categories:

Function(s) Process(es)

methylation
mitochondrial respiratory chain complex I assembly
molecular_function
methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0