This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00917735_m1
See other CDH7 GE Assays ›
Gene Symbol:
CDH7
Gene Name:
cadherin 7
Gene Aliases:
CDH7L1
Chromosome Location:
Chr.18: 65750252 - 65885136 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00917735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317214.1 NP_001304143.1 8 - 9 2063 109
NM_004361.3 NP_004352.2 8 - 9 2063 109
NM_033646.2 NP_387450.1 8 - 9 1693 109
XM_017025523.1 XP_016881012.1 8 - 9 2379 109
GenBank mRNA AB035301.1 - 8 - 9 1794 109
AJ007611.1 - 8 - 9 1691 109
BC036786.1 - 8 - 9 2063 5744842 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800007570.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000235.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH7

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
single organismal cell-cell adhesion
adherens junction organization
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000581601.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 ENST00000581601.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 ENST00000323011.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 ENST00000397966.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 ENST00000397968.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 ENST00000536984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 NM_004361.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 NM_033646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002798.hg.1 TC18000235.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000581601.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 ENST00000581601.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay TC1800007570.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 NM_033646.2 Refseq
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 TC1800007570.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 ENST00000323011.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 ENST00000397968.3 Ensembl
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 ENST00000536984.6 Ensembl
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 NM_004361.3 Refseq
Affymetrix Human Clariom D Assay PSR1800152014.hg.1 NM_033646.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline