This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00918014_m1
See other IKZF3 GE Assays ›
Gene Symbol:
IKZF3
Gene Name:
IKAROS family zinc finger 3
Gene Aliases:
AIO, AIOLOS, ZNFN1A3
Chromosome Location:
Chr.17: 39757715 - 39864188 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00918014_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IKZF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257410.1 NP_001244339.1 2 - 3 125 95
NM_001257411.1 NP_001244340.1 2 - 3 125 95
NM_001257412.1 NP_001244341.1 2 - 3 125 95
NM_001257413.1 NP_001244342.1 2 - 3 125 95
NM_001257414.1 NP_001244343.1 2 - 3 125 95
NM_001284514.1 NP_001271443.1 2 - 3 125 95
NM_001284515.1 NP_001271444.1 2 - 3 125 95
NM_012481.4 NP_036613.2 2 - 3 125 95
NM_183228.2 NP_899051.1 2 - 3 125 95
NM_183229.2 NP_899052.1 2 - 3 125 95
NM_183230.2 NP_899053.1 2 - 3 125 95
NM_183231.2 NP_899054.1 2 - 3 125 95
NM_183232.2 NP_899055.1 2 - 3 125 95
GenBank mRNA AF129512.1 - 2 - 3 125 95
AJ292565.1 - 1 - 2 63 95
AJ292566.1 - 1 - 2 63 95
AJ292567.1 - 1 - 2 63 95
AJ292568.1 - 1 - 2 63 95
AJ292569.1 - 1 - 2 63 95
AY377976.1 - 2 - 3 125 95
AY377977.1 - 2 - 3 125 95
AY377978.1 - 2 - 3 125 95
AY377979.1 - 2 - 3 125 95
AY377980.1 - 2 - 3 125 95
AY377981.1 - 2 - 3 125 95
AY377982.1 - 2 - 3 125 95
BC032707.1 - 2 - 3 114 5497068 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700010587.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001284514.1,NM_001284515.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IKZF3

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
mesoderm development
regulation of B cell proliferation
B cell activation
regulation of apoptotic process
regulation of B cell differentiation
regulation of lymphocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000293068.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000346243.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000346872.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000348427.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000350532.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000351680.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377944.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377945.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377952.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377958.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000394189.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000439016.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000467757.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000623724.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257410.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257411.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257412.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257413.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257414.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001284514.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001284515.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_012481.4 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183228.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183229.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183230.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183231.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183232.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000293068.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000346243.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000346872.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000348427.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000350532.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000351680.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377944.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377945.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377952.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000377958.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000394189.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000439016.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000467757.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 ENST00000623724.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257410.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257411.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257412.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257413.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001257414.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001284514.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_001284515.1 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_012481.4 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183228.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183229.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183230.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183231.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 NM_183232.2 Refseq
Affymetrix Human Clariom D Assay PSR1700183436.hg.1 TC1700010587.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0