This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00918105_g1
See other OBSCN GE Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00918105_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OBSCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 93 - 94 20320 57
NM_001271223.2 NP_001258152.2 104 - 105 23191 57
XM_005273287.4 XP_005273344.1 105 - 106 23577 57
XM_005273291.4 XP_005273348.1 105 - 106 23541 57
XM_005273298.4 XP_005273355.1 104 - 105 23268 57
XM_005273307.4 XP_005273364.1 100 - 101 22455 57
XM_006711822.3 XP_006711885.1 103 - 104 23001 57
XM_006711823.3 XP_006711886.1 103 - 104 22995 57
XM_011544297.2 XP_011542599.1 97 - 98 21411 57
XM_011544299.2 XP_011542601.1 96 - 97 21135 57
XM_017002443.1 XP_016857932.1 105 - 106 23622 57
XM_017002444.1 XP_016857933.1 105 - 106 23585 57
XM_017002445.1 XP_016857934.1 105 - 106 23619 57
XM_017002446.1 XP_016857935.1 105 - 106 23619 57
XM_017002447.1 XP_016857936.1 105 - 106 23619 57
XM_017002448.1 XP_016857937.1 105 - 106 23472 57
XM_017002449.1 XP_016857938.1 104 - 105 23359 57
XM_017002450.1 XP_016857939.1 104 - 105 23359 57
XM_017002451.1 XP_016857940.1 104 - 105 23359 57
XM_017002452.1 XP_016857941.1 104 - 105 23359 57
XM_017002453.1 XP_016857942.1 104 - 105 23346 57
XM_017002454.1 XP_016857943.1 104 - 105 23346 57
XM_017002455.1 XP_016857944.1 104 - 105 23346 57
XM_017002456.1 XP_016857945.1 104 - 105 23346 57
XM_017002457.1 XP_016857946.1 104 - 105 23346 57
XM_017002458.1 XP_016857947.1 103 - 104 23095 57
XM_017002459.1 XP_016857948.1 103 - 104 23095 57
XM_017002460.1 XP_016857949.1 103 - 104 23095 57
XM_017002461.1 XP_016857950.1 103 - 104 23082 57
XM_017002462.1 XP_016857951.1 103 - 104 23070 57
XM_017002463.1 XP_016857952.1 103 - 104 23049 57
XM_017002464.1 XP_016857953.1 103 - 104 22998 57
XM_017002465.1 XP_016857954.1 102 - 103 22831 57
XM_017002466.1 XP_016857955.1 102 - 103 22831 57
XM_017002467.1 XP_016857956.1 102 - 103 22831 57
XM_017002468.1 XP_016857957.1 101 - 102 22567 57
XM_017002469.1 XP_016857958.1 99 - 100 22039 57
XM_017002470.1 XP_016857959.1 94 - 95 20720 57
GenBank mRNA AB046859.2 - 33 - 34 4095 57
AM231061.1 - 19 - 20 2222 57
DQ400343.1 - 19 - 20 2222 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011861.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001888.hg.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01029492.hg.1 TC01001888.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01029492.hg.1 ENST00000366707.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029492.hg.1 ENST00000422127.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029492.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187654.hg.1 TC0100011861.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0