This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00918210_m1
See other PPP2R3B GE Assays ›
Gene Symbol:
PPP2R3B
Gene Name:
protein phosphatase 2 regulatory subunit B''beta
Gene Aliases:
NYREN8, PPP2R3L, PPP2R3LY, PR48
Chromosome Location:
Chr.Y: 333933 - 386955 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00918210_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013239.4 NP_037371.2 10 - 11 1611 72
XM_011545630.2 XP_011543932.1 10 - 11 1611 72
GenBank mRNA AB209137.1 - 6 - 7 1217 72
AF135016.1 - 9 - 10 975 72
AF155098.1 - 4 - 5 386 72
AL157501.1 - 2 - 3 322 72
BC009032.1 - 2 - 3 568 72
BC011180.1 - 4 - 5 388 72
BC063429.1 - 10 - 11 1366 5239974 72
BM715684.1 - 4 - 5 351 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008888.hg.1, TC0Y00006863.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000795.hg.1, TC0Y000112.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP2R3B

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
cell cycle arrest
regulation of protein phosphatase type 2A activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
calcium ion binding
protein binding
protein phosphatase type 2A regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X010853.hg.1 ENST00000390665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X010853.hg.1 NM_013239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X010853.hg.1 TC0X000795.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0Y001247.hg.1 ENST00000390665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0Y001247.hg.1 NM_013239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0Y001247.hg.1 TC0Y000112.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000390665.7 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000468169.5 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000477110.5 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000479438.5 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 NM_013239.4 Refseq
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000390665.7 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000468169.5 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000477110.5 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 ENSTR0000479438.5 Ensembl
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 NM_013239.4 Refseq
Affymetrix Human Clariom D Assay PSR0Y00146731.hg.1 TC0Y00006863.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000390665.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000468169.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000477110.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000479438.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000390665.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000468169.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000477110.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 ENST00000479438.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00161094.hg.1 TC0X00008888.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-bxd7r:80/100.66.79.78:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline