This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00918343_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs00918343_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ437370.1 - 1 111 Chr. X - 71054905 - 71055644 [-] on Build GRCh38 136
GenBankmRNA DB324115.1 - 1 Chr. X - 71055158 - 71055629 [+] on Build GRCh38 136
GenBankmRNA AK023112.1 - 1 1622 Chr. X - 71054664 - 71057155 [-] on Build GRCh38 136
GenBankmRNA BM745450.1 - 1 235 Chr. X - 71055257 - 71055768 [-] on Build GRCh38 136
GenBankmRNA BE671426.1 - 2 Chr. X - 70993014 - 71055803 [+] on Build GRCh38 136

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 7 member 3 SLC7A3 ATRC3,
CAT-3,
CAT3
-124.0kb Anti-sense Chr. X - 70925579 - 70931125 on NCBI Build GRCh38
Downstream sorting nexin 12 SNX12 1.1kb Anti-sense Chr. X - 71059247 - 71073426 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline