This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00918564_g1
See other DMTN GE Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00918564_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DMTN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114135.4 NP_001107607.1 12 - 13 1225 54
NM_001114136.2 NP_001107608.1 12 - 13 1181 54
NM_001302817.2 NP_001289746.1 10 - 11 1194 54
NM_001323378.1 NP_001310307.1 12 - 13 1314 54
NM_001323380.1 NP_001310309.1 12 - 13 1221 54
NM_001323381.1 NP_001310310.1 11 - 12 1330 54
NM_001323382.1 NP_001310311.1 12 - 13 1260 54
NM_001323388.1 NP_001310317.1 10 - 11 1061 54
NM_001323389.1 NP_001310318.1 11 - 12 1269 54
NM_001323390.1 NP_001310319.1 11 - 12 1136 54
NM_001323391.1 NP_001310320.1 11 - 12 1239 54
NM_001323392.1 NP_001310321.1 11 - 12 1400 54
NM_001323393.1 NP_001310322.1 11 - 12 1106 54
NM_001323394.1 NP_001310323.1 11 - 12 1267 54
NM_001323395.1 NP_001310324.1 11 - 12 1146 54
NM_001323396.1 NP_001310325.1 11 - 12 1150 54
NM_001323397.1 NP_001310326.1 11 - 12 1318 54
NM_001978.4 NP_001969.2 12 - 13 1475 54
XM_005273432.1 XP_005273489.1 12 - 13 1207 54
XM_005273433.1 XP_005273490.1 12 - 13 1139 54
XM_005273442.1 XP_005273499.1 11 - 12 991 54
XM_011544435.1 XP_011542737.1 11 - 12 1400 54
XM_017013193.1 XP_016868682.1 10 - 11 1167 54
XM_017013194.1 XP_016868683.1 10 - 11 1034 54
GenBank mRNA AB209718.1 - 8 - 9 701 54
AK289650.1 - 12 - 13 1107 54
AK295452.1 - 10 - 11 1080 54
BC006318.2 - 12 - 13 1224 4125804 54
BC017445.1 - 12 - 13 1224 3162732 54
BT007396.1 - 11 - 12 952 54
U28389.1 - 12 - 13 1402 54

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DMTN

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0