This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00919305_s1
See other PRMT2 GE Assays ›
Gene Symbol:
PRMT2
Gene Name:
protein arginine methyltransferase 2
Gene Aliases:
HRMT1L1
Chromosome Location:
Chr.21: 46635153 - 46665685 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00919305_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRMT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001242865.2 NP_001229794.1 7 2504 91
XM_006723999.2 XP_006724062.1 8 1574 91
GenBank mRNA AK055950.1 - 1 240 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA682776.1 - 1 240 Chr. 21 - 46653191 - 46653971 [+] on Build GRCh38 91
GenBankmRNA DR003200.1 - 1 451 Chr. 21 - 46652985 - 46653505 [+] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream S100 calcium binding protein B S100B NEF,
S100,
S100-B,
S100beta
-46.7kb Anti-sense Chr. 21 - 46598618 - 46605242 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007474.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000265.hg.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
signal transduction
histone methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
negative regulation of NF-kappaB transcription factor activity
histone arginine methylation
positive regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
developmental cell growth
regulation of androgen receptor signaling pathway
negative regulation of G1/S transition of mitotic cell cycle
transcription coactivator activity
signal transducer activity
protein binding
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
estrogen receptor binding
progesterone receptor binding
protein-arginine omega-N asymmetric methyltransferase activity
histone methyltransferase activity
protein homodimerization activity
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
thyroid hormone receptor binding
androgen receptor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21003330.hg.1 NM_001242865.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21003330.hg.1 TC21000265.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR21003331.hg.1 NM_001242865.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21003331.hg.1 TC21000265.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100008480.hg.1 ENST00000419906.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100150839.hg.1 NM_001242865.2 Refseq
Affymetrix Human Clariom D Assay PSR2100150839.hg.1 TC2100007474.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100150839.hg.1 NM_001242865.2 Refseq
Affymetrix Human Clariom D Assay PSR2100150838.hg.1 ENST00000455177.3 Ensembl
Affymetrix Human Clariom D Assay PSR2100150838.hg.1 NM_001242865.2 Refseq
Affymetrix Human Clariom D Assay PSR2100150838.hg.1 TC2100007474.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100150838.hg.1 ENST00000455177.3 Ensembl
Affymetrix Human Clariom D Assay PSR2100150838.hg.1 NM_001242865.2 Refseq
Affymetrix Human Clariom D Assay TC2100008480.hg.1 ENST00000419906.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 705157 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0