This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00920067_m1
See other NDUFAF5 GE Assays ›
Gene Symbol:
NDUFAF5
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases:
C20orf7, bA526K24.2, dJ842G6.1
Chromosome Location:
Chr.20: 13785026 - 13821580 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00920067_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFAF5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039375.2 NP_001034464.1 5 - 6 476 187
NM_024120.4 NP_077025.2 6 - 7 560 271
NR_029377.1 6 - 7 560 271
XM_006723620.3 XP_006723683.1 6 - 7 537 271
XM_006723622.2 XP_006723685.1 5 - 6 441 167
XM_006723623.2 XP_006723686.1 5 - 6 573 187
XM_006723624.2 XP_006723687.1 5 - 6 587 167
XM_011529342.2 XP_011527644.1 6 - 7 537 271
XM_017028061.1 XP_016883550.1 6 - 7 672 167
GenBank mRNA AK091060.1 - 6 - 7 537 271
AK289781.1 - 6 - 7 552 271
BC005984.1 - 6 - 7 370 271
BC073158.1 - 5 - 6 453 6053315 187
CB156360.1 - 5 - 6 446 167
DA289766.1 - 5 - 6 476 187

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC005984.1, XM_006723622.2, XM_006723623.2, XM_006723624.2, XM_017028061.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFAF5

Gene Ontology Categories:

Function(s) Process(es)

methylation
mitochondrial respiratory chain complex I assembly
molecular_function
methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0