Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020746.4 | NP_065797.2 | 3 - 4 | 463 | 94 | |
| GenBank mRNA | AB033097.1 | - | 2 - 3 | 369 | 94 | |
| AB097003.1 | - | 3 - 4 | 426 | 94 | ||
| AB232371.1 | - | 2 - 3 | 363 | 94 | ||
| AK023799.1 | - | 3 - 4 | 419 | 94 | ||
| BC044952.1 | - | 3 - 4 | 397 | 5751684 | 94 | |
| DB219666.1 | - | 3 - 4 | 463 | 94 | ||
| DQ167126.1 | - | 2 - 3 | 291 | 94 | ||
| DQ174270.1 | - | 2 - 3 | 291 | 94 | ||
| DQ181928.1 | - | 2 - 3 | 291 | 94 | ||
| KC415005.1 | - | 2 - 3 | 291 | 94 | ||
More Information
Set Membership: |
|

MAVS
Genomic Map