This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00920261_m1
See other PPP3CB GE Assays ›
Gene Symbol:
PPP3CB
Gene Name:
protein phosphatase 3 catalytic subunit beta
Gene Aliases:
CALNA2, CALNB, CNA2, PP2Bbeta
Chromosome Location:
Chr.10: 73436428 - 73496024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00920261_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP3CB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289968.1 NP_001276897.1 3 - 4 550 154
GenBank mRNA M29550.1 - 3 - 4 522 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011025.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of T cell mediated cytotoxicity
lymphangiogenesis
protein phosphorylation
protein dephosphorylation
signal transduction
Wnt signaling pathway, calcium modulating pathway
heart development
learning
memory
dephosphorylation
calcium ion regulated exocytosis
T cell differentiation
locomotion involved in locomotory behavior
calcineurin-NFAT signaling cascade
response to cytokine
social behavior
cellular response to drug
positive regulation of insulin secretion involved in cellular response to glucose stimulus
Fc-epsilon receptor signaling pathway
T cell proliferation
T cell activation
T cell homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of synaptic plasticity
axon extension
regulation of insulin secretion
positive regulation of NFAT protein import into nucleus
protein serine/threonine phosphatase activity
calcium ion binding
protein binding
calmodulin binding
drug binding
enzyme binding
protein phosphatase 2B binding
calmodulin-dependent protein phosphatase activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1000175091.hg.1 NM_001289968.1 Refseq
Affymetrix Human Clariom D Assay PSR1000175091.hg.1 NM_001289968.1 Refseq
Affymetrix Human Clariom D Assay PSR1000175091.hg.1 TC1000011025.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0