This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00920847_g1
See other ACRV1 GE Assays ›
Gene Symbol:
ACRV1
Gene Name:
acrosomal vesicle protein 1
Gene Aliases:
D11S4365, SP-10, SPACA2
Chromosome Location:
Chr.11: 125672334 - 125680898 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00920847_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACRV1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001612.5 NP_001603.1 2 615 198
NM_020107.4 NP_064492.1 3 450 198
NM_020108.4 NP_064493.1 3 405 198
GenBank mRNA AK223335.1 - 2 564 198
BC014588.2 - 2 591 4825974 198
BG720361.1 - 3 375 198
BI826178.1 - 3 383 198
M82968.1 - 2 549 198

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012712.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002422.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ACRV1

Gene Ontology Categories:

Function(s)

multicellular organism development

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000257382.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000345274.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000348856.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000426183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000445562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000530048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 ENST00000533904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 NM_001612.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 NM_020107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 NM_020108.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11029036.hg.1 TC11002422.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000315608.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020069.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay TC1100012712.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 ENST00000527795.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 ENST00000530048.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 ENST00000533904.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 NM_001612.5 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 NM_020107.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 NM_020108.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197750.hg.1 TC1100012712.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline