This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00920922_m1
See other TMPRSS13 GE Assays ›
Gene Symbol:
TMPRSS13
Gene Name:
transmembrane protease, serine 13
Gene Aliases:
MSP, MSPL, MSPS, TMPRSS11
Chromosome Location:
Chr.11: 117900641 - 117929453 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00920922_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077263.2 NP_001070731.1 3 - 4 700 72
NM_001206790.1 NP_001193719.1 3 - 4 700 72
NM_001244995.1 NP_001231924.1 3 - 4 700 72
GenBank mRNA AK027798.1 - 3 - 4 644 72
AK300283.1 - 3 - 4 612 72
BC018715.1 - 4 - 5 435 72
BC114928.1 - 3 - 4 619 40028749 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
receptor-mediated endocytosis
serine-type endopeptidase activity
scavenger receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline