This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00921937_m1
See other SLC7A11 GE Assays ›
Gene Symbol:
SLC7A11
Gene Name:
solute carrier family 7 member 11
Gene Aliases:
CCBR1, xCT
Chromosome Location:
Chr.4: 138164094 - 138242418 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00921937_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014331.3 NP_055146.1 6 - 7 1064 93
XM_011531802.2 XP_011530104.1 6 - 7 1133 93
GenBank mRNA AB026891.1 - 6 - 7 1019 93
AB040875.1 - 6 - 7 919 93
AF200708.1 - 6 - 7 1029 93
AF252872.1 - 6 - 7 1015 93
AJ277882.1 - 6 - 7 784 93
AK290359.1 - 6 - 7 1064 93
AK314855.1 - 6 - 7 958 93
BC012087.1 - 6 - 7 944 4562994 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011920.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001570.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A11

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transport
response to oxidative stress
brain development
response to toxic substance
response to nicotine
leukocyte migration
lens fiber cell differentiation
platelet aggregation
L-glutamate transmembrane transport
protein binding
cystine:glutamate antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04021971.hg.1 TC04001570.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04021971.hg.1 ENST00000280612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021971.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 ENST00000509248.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay TC0400011920.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay PSR0400181829.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400181829.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay PSR0400181829.hg.1 ENST00000280612.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400181829.hg.1 NM_014331.3 Refseq
Affymetrix Human Clariom D Assay PSR0400181829.hg.1 TC0400011920.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0