This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00922215_g1
See other CEACAM3 GE Assays ›
Gene Symbol:
CEACAM3
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 3
Gene Aliases:
CD66D, CEA, CGM1, W264, W282
Chromosome Location:
Chr.19: 41796590 - 41811553 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00922215_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277163.2 NP_001264092.1 4 - 5 658 127
GenBank mRNA L00693.1 - 4 - 5 624 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008161.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000344550.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000357396.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000415495.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000630848.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 NM_001277163.2 Refseq
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 NM_001815.4 Refseq
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 NR_102333.2 Refseq
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 TC1900008161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000344550.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000357396.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000415495.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 ENST00000630848.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 NM_001277163.2 Refseq
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 NM_001815.4 Refseq
Affymetrix Human Clariom D Assay PSR1900162126.hg.1 NR_102333.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0