This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00922568_m1
See other PIGR GE Assays ›
Gene Symbol:
PIGR
Gene Name:
polymeric immunoglobulin receptor
Gene Aliases:
-
Chromosome Location:
Chr.1: 206928522 - 206946466 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00922568_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIGR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002644.3 NP_002635.2 8 - 9 2197 82
XM_011509629.1 XP_011507931.1 8 - 9 2108 82
GenBank mRNA AF272149.2 - 8 - 9 2193 82
BC110494.2 - 8 - 9 2094 40007650 82
BC110495.2 - 8 - 9 2094 40007652 82
CR749533.1 - 9 - 10 2270 82
M24559.1 - 6 - 7 1800 82
S62403.1 - 8 - 9 2169 82
X73079.1 - 8 - 9 2193 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100017113.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003768.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PIGR

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
retina homeostasis
immunoglobulin transcytosis in epithelial cells mediated by polymeric immunoglobulin receptor
epidermal growth factor receptor signaling pathway
Fc receptor signaling pathway
receptor clustering
polymeric immunoglobulin receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01057776.hg.1 TC01003768.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01057776.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01057776.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01057776.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000491503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 ENST00000491503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay PSR0100229753.hg.1 TC0100017113.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000356495.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000487208.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000491503.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 ENST00000491503.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq
Affymetrix Human Clariom D Assay TC0100017113.hg.1 NM_002644.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0