This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00922999_m1
See other PNLIPRP3 GE Assays ›
Gene Symbol:
PNLIPRP3
Gene Name:
pancreatic lipase related protein 3
Gene Aliases:
-
Chromosome Location:
Chr.10: 116427879 - 116477961 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00922999_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNLIPRP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011709.2 NP_001011709.2 2 - 3 303 111
XM_011539276.1 XP_011537578.1 2 - 3 336 111
GenBank mRNA AL833418.1 - 2 - 3 352 111
BC117224.1 - 2 - 3 303 40125775 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009002.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000841.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PNLIPRP3

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
triglyceride lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10010181.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010181.hg.1 ENST00000481306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010181.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010181.hg.1 TC10000841.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010178.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010178.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010178.hg.1 TC10000841.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10010179.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010179.hg.1 ENST00000481306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010179.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010179.hg.1 TC10000841.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10010177.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010177.hg.1 NM_001011709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010177.hg.1 TC10000841.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000161610.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161610.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161610.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161610.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161610.hg.1 TC1000009002.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000161609.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161609.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161609.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161609.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161609.hg.1 TC1000009002.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000161608.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161608.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161608.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161608.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161608.hg.1 TC1000009002.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay TC1000009002.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161611.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161611.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161611.hg.1 ENST00000369230.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000161611.hg.1 NM_001011709.2 Refseq
Affymetrix Human Clariom D Assay PSR1000161611.hg.1 TC1000009002.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0