This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00923606_g1
See other ZNF268 GE Assays ›
Gene Symbol:
ZNF268
Gene Name:
zinc finger protein 268
Gene Aliases:
HZF3
Chromosome Location:
Chr.12: 133181409 - 133207112 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00923606_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF268
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165881.2 NP_001159353.1 4 - 5 571 98
NM_001165882.2 NP_001159354.1 4 - 5 578 98
NM_001165887.1 NP_001159359.1 3 - 4 494 98
NM_003415.2 NP_003406.1 4 - 5 695 98
NM_152943.2 NP_694422.2 4 - 5 695 98
GenBank mRNA AF317549.1 - 4 - 5 695 98
AF385187.1 - 4 - 5 545 98
AK027712.1 - 4 - 5 609 98
AY583691.1 - 4 - 5 481 98
BC110542.2 - 4 - 5 571 40036577 98
CN255799.1 - 4 - 5 555 98
DQ057360.1 - 3 - 4 283 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF385187.1, AK027712.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZNF268

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of mitotic cell cycle
positive regulation of cell proliferation
negative regulation of cell proliferation
cell differentiation
positive regulation of cell migration
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter during mitosis
negative regulation of cell cycle arrest
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0