This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00925923_g1
See other MIOX GE Assays ›
Gene Symbol:
MIOX
Gene Name:
myo-inositol oxygenase
Gene Aliases:
ALDRL6
Chromosome Location:
Chr.22: 50483988 - 50490321 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00925923_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIOX
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017584.5 NP_060054.4 1 - 2 131 68
XM_011530705.1 XP_011529007.1 2 - 3 2481 68
GenBank mRNA AY064416.1 - 1 - 2 44 68
BC012025.1 - 1 - 2 131 4454512 68
BC064395.1 - 1 - 2 39 68
BC073848.1 - 1 - 2 51 5186354 68
CR456478.1 - 1 - 2 76 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200007809.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000427.hg.1

Additional Information:

This assay is in the 5'UTR region of the AY064416.1,BC012025.1,BC073848.1,CR456478.1,NM_017584.5,XM_011530705.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MIOX

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

inositol catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
ferric iron binding
oxidoreductase activity, acting on NAD(P)H
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen
inositol oxygenase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22008087.hg.1 ENST00000216075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008087.hg.1 ENST00000395732.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008087.hg.1 ENST00000395733.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008087.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008087.hg.1 TC22000427.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22008088.hg.1 ENST00000216075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008088.hg.1 ENST00000395732.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008088.hg.1 ENST00000395733.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008088.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008088.hg.1 TC22000427.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008084.hg.1 ENST00000395733.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008084.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008084.hg.1 TC22000427.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22008085.hg.1 ENST00000216075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008085.hg.1 ENST00000395733.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008085.hg.1 NM_017584.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008085.hg.1 TC22000427.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0800011024.hg.1 ENST00000521205.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 TC2200007809.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200155460.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC0800011024.hg.1 ENST00000521205.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 TC2200007809.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155456.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay PSR2200155455.hg.1 TC2200007809.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200155455.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155455.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay PSR2200155455.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200155455.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000216075.10 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395732.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000395733.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 ENST00000451761.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Affymetrix Human Clariom D Assay TC2200007809.hg.1 NM_017584.5 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 167768 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0