This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00929130_m1
See other ECHS1 GE Assays ›
Gene Symbol:
ECHS1
Gene Name:
enoyl-CoA hydratase, short chain 1
Gene Aliases:
ECHS1D, SCEH
Chromosome Location:
Chr.10: 133362483 - 133373404 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00929130_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECHS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004092.3 NP_004083.3 5 - 6 689 106
GenBank mRNA BC008906.2 - 5 - 6 632 2907059 106
BC013264.1 - 5 - 6 624 106
BQ130816.1 - 3 - 4 324 106
BT007123.1 - 5 - 6 618 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012399.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001790.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ECHS1

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
enoyl-CoA hydratase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10023639.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10023639.hg.1 NM_004092.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10023639.hg.1 TC10001790.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay TC1000012399.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay PSR1000184649.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000184649.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay PSR1000184649.hg.1 ENST00000368547.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000184649.hg.1 NM_004092.3 Refseq
Affymetrix Human Clariom D Assay PSR1000184649.hg.1 TC1000012399.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0