This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00929568_g1
See other PPP1R1A GE Assays ›
Gene Symbol:
PPP1R1A
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 1A
Gene Aliases:
I1, IPP1
Chromosome Location:
Chr.12: 54579240 - 54588659 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00929568_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006741.3 NP_006732.3 4 - 5 416 69
GenBank mRNA AY063766.1 - 4 - 5 247 69
BC022470.1 - 4 - 5 416 4819907 69
BP211831.1 - 4 - 5 416 69
CR457003.1 - 4 - 5 245 69
U48707.1 - 4 - 5 253 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200010842.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001574.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R1A

Panther Classification:

Molecular Function -

phosphatase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

glycogen metabolic process
negative regulation of protein kinase activity
intracellular signal transduction
protein serine/threonine phosphatase inhibitor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12020870.hg.1 ENST00000257905.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12020870.hg.1 NM_006741.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12020870.hg.1 TC12001574.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay PSR1200183342.hg.1 TC1200010842.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000257905.12 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547431.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000547826.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 ENST00000553113.5 Ensembl
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq
Affymetrix Human Clariom D Assay TC1200010842.hg.1 NM_006741.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0