This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00930429_m1
See other GLYATL2 GE Assays ›
Gene Symbol:
GLYATL2
Gene Name:
glycine-N-acyltransferase like 2
Gene Aliases:
BXMAS2-10, GATF-B
Chromosome Location:
Chr.11: 58834065 - 58905699 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00930429_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLYATL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145016.3 NP_659453.3 1 - 2 92 83
GenBank mRNA AF426250.1 - 1 - 2 352 83
BC021682.1 - 1 - 2 79 4079236 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100010952.hg
This assay targets the same location as HTA transcript cluster(s) TC11001814.hg

Additional Information:

This assay is in the 5'UTR region of the AF426250.1,BC021682.1,NM_145016.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLYATL2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Process(es)

glycine N-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11020975.hg.1 ENST00000287275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11020975.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11020975.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11020975.hg.1 TC11001814.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11020973.hg.1 ENST00000287275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11020973.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11020973.hg.1 NM_145016.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11020973.hg.1 TC11001814.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182672.hg.1 TC1100010952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182674.hg.1 TC1100010952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 ENST00000533636.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay TC1100010952.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 ENST00000287275.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 ENST00000532258.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 NM_145016.3 Refseq
Affymetrix Human Clariom D Assay PSR1100182671.hg.1 TC1100010952.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0