This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00931449_m1
See other SLC7A1 GE Assays ›
Gene Symbol:
SLC7A1
Gene Name:
solute carrier family 7 member 1
Gene Aliases:
ATRC1, CAT-1, ERR, HCAT1, REC1L
Chromosome Location:
Chr.13: 29509410 - 29595715 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00931449_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003045.4 NP_003036.1 11 - 12 2066 75
XM_005266507.3 XP_005266564.1 11 - 12 1861 75
XM_017020716.1 XP_016876205.1 11 - 12 2077 75
GenBank mRNA AB105377.1 - 11 - 12 1953 75
AF078107.1 - 10 - 11 1799 75
AK312818.1 - 11 - 12 1840 75
BC063303.1 - 11 - 12 1863 30340706 75
BC069358.1 - 10 - 11 1748 7262451 75
BC115407.1 - 10 - 11 1748 40076678 75
X57303.1 - 11 - 12 1829 75
X59155.1 - 11 - 12 1826 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008487.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000523.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A1

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
amino acid transport
arginine transport
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
L-ornithine transmembrane transporter activity
protein binding
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005918.hg.1 ENST00000380752.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005918.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005918.hg.1 TC13000523.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13005920.hg.1 ENST00000380752.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005920.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005920.hg.1 NM_003045.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005920.hg.1 TC13000523.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000450494.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay TC1300008487.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay PSR1300157213.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay PSR1300157213.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay PSR1300157213.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay PSR1300157213.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay PSR1300157213.hg.1 TC1300008487.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 ENST00000380752.9 Ensembl
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 ENST00000473577.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 NM_003045.4 Refseq
Affymetrix Human Clariom D Assay PSR1300157215.hg.1 TC1300008487.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0