This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00931908_g1
See other IL18BP GE Assays ›
Gene Symbol:
IL18BP
Gene Name:
interleukin 18 binding protein
Gene Aliases:
IL18BPa
Chromosome Location:
Chr.11: 71997944 - 72002919 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00931908_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL18BP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039659.1 NP_001034748.1 2 - 3 221 82
GenBank mRNA BC044215.1 - 2 - 3 203 5755934 82
BX357194.2 - 2 - 3 241 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000747.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC044215.1,NM_001039659.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to lipopolysaccharide
T-helper 1 type immune response
cellular response to hydrogen peroxide
cellular response to tumor necrosis factor
extracellular negative regulation of signal transduction
interleukin-18 binding
receptor antagonist activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000260049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000337131.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000393703.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000393705.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000393707.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000404792.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000497194.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 ENST00000531053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_001039659.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_001039660.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_001145055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_001145057.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_005699.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_173042.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 NM_173044.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11009705.hg.1 TC11000747.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0