This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00932077_m1
See other RBM26 GE Assays ›
Gene Symbol:
RBM26
Gene Name:
RNA binding motif protein 26
Gene Aliases:
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location:
Chr.13: 79311827 - 79406265 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00932077_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286632.1 NP_001273561.1 12 - 13 2306 83
XM_005266491.4 XP_005266548.1 12 - 13 2388 83
XM_005266497.1 XP_005266554.1 12 - 13 2388 83
XM_006719857.1 XP_006719920.1 12 - 13 2388 83
XM_011535179.1 XP_011533481.1 12 - 13 2388 83
XM_011535180.2 XP_011533482.1 12 - 13 2388 83
XM_011535182.1 XP_011533484.1 12 - 13 2373 83
XM_011535184.1 XP_011533486.1 12 - 13 2358 83
XM_011535185.2 XP_011533487.1 12 - 13 2358 83
XM_011535186.2 XP_011533488.1 12 - 13 2350 83
XM_011535188.2 XP_011533490.1 12 - 13 2350 83
XM_011535189.1 XP_011533491.1 12 - 13 2388 83
XM_011535192.2 XP_011533494.1 12 - 13 2350 83
XM_011535195.1 XP_011533497.1 12 - 13 2086 83
XM_017020688.1 XP_016876177.1 12 - 13 2388 83
XM_017020690.1 XP_016876179.1 12 - 13 2380 83
XM_017020693.1 XP_016876182.1 12 - 13 2388 83
XM_017020694.1 XP_016876183.1 12 - 13 2373 83
XM_017020696.1 XP_016876185.1 12 - 13 2380 83
XM_017020697.1 XP_016876186.1 12 - 13 2380 83
XM_017020698.1 XP_016876187.1 12 - 13 2343 83
XM_017020699.1 XP_016876188.1 12 - 13 2365 83
XM_017020701.1 XP_016876190.1 12 - 13 2388 83
XM_017020702.1 XP_016876191.1 12 - 13 2388 83
XM_017020703.1 XP_016876192.1 12 - 13 2358 83
GenBank mRNA AK027456.1 - 6 - 7 955 83
AK302871.1 - 12 - 13 1915 83
AL834343.1 - 7 - 8 1219 83
AX775885.1 - 6 - 7 955 83
BC111697.1 - 12 - 13 1867 40038080 83
BX648672.1 - 12 - 13 1864 83

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
negative regulation of phosphatase activity
nucleotide binding
protein binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000267229.11 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000438724.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000438737.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000622611.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 NM_001286631.1 Refseq
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 NM_001286632.1 Refseq
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 NM_022118.4 Refseq
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000267229.11 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000438724.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000438737.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 ENST00000622611.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 NM_001286631.1 Refseq
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 NM_001286632.1 Refseq
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 NM_022118.4 Refseq
Affymetrix Human Clariom D Assay PSR1300162647.hg.1 TC1300009330.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0