This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00932433_g1
See other PCBD2 GE Assays ›
Gene Symbol:
PCBD2
Gene Name:
pterin-4 alpha-carbinolamine dehydratase 2
Gene Aliases:
DCOH2, DCOHM, PHS2
Chromosome Location:
Chr.5: 134905120 - 134962646 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00932433_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCBD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032151.4 NP_115527.3 2 - 3 234 94
GenBank mRNA AF499009.1 - 2 - 3 154 94
AK098495.1 - 2 - 3 206 94
AL136721.1 - 2 - 3 234 94
BC054021.1 - 2 - 3 237 6385036 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500013230.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05003413.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PCBD2

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

tetrahydrobiopterin biosynthetic process
positive regulation of transcription, DNA-templated
protein homotetramerization
protein heterooligomerization
oxidation-reduction process
phenylalanine 4-monooxygenase activity
protein binding
4-alpha-hydroxytetrahydrobiopterin dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05009356.hg.1 TC05003413.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05009356.hg.1 ENST00000254908.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05009356.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05009356.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009485.hg.1 ENST00000575326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009485.hg.1 ENST00000575326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC0500013230.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay TC1700009485.hg.1 ENST00000575326.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009485.hg.1 ENST00000575326.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 ENST00000254908.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 ENST00000510013.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 NM_032151.4 Refseq
Affymetrix Human Clariom D Assay PSR0500194156.hg.1 TC0500013230.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0