This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00934060_m1
See other LIG4 GE Assays ›
Gene Symbol:
LIG4
Gene Name:
DNA ligase 4
Gene Aliases:
LIG4S
Chromosome Location:
Chr.13: 108207442 - 108218368 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00934060_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIG4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002312.3 NP_002303.2 1 - 2 245 111
NM_206937.1 NP_996820.1 2 - 3 124 111
XM_005254057.4 XP_005254114.1 2 - 3 259 111
XM_006719951.3 XP_006720014.1 2 - 3 1964 111
XM_011521091.2 XP_011519393.1 3 - 4 586 111
XM_011521092.2 XP_011519394.1 2 - 3 162 111
XM_017020569.1 XP_016876058.1 3 - 4 2227 111
XM_017020570.1 XP_016876059.1 3 - 4 506 111
GenBank mRNA BC037491.2 - 2 - 3 124 5259632 111
X83441.1 - 1 - 2 245 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009740.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000867.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC037491.2,NM_002312.3,NM_206937.1,X83441.1,XM_005254057.4,XM_006719951.3,XM_011521091.2,XM_011521092.2,XM_017020569.1,XM_017020570.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LIG4

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

single strand break repair
in utero embryonic development
pro-B cell differentiation
DNA ligation
lagging strand elongation
nucleotide-excision repair, DNA gap filling
double-strand break repair
double-strand break repair via nonhomologous end joining
cell cycle
central nervous system development
cell proliferation
response to X-ray
response to gamma radiation
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
T cell receptor V(D)J recombination
somatic stem cell population maintenance
negative regulation of neuron apoptotic process
isotype switching
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
DNA ligation involved in DNA recombination
DNA ligation involved in DNA repair
chromosome organization
cell division
neuron apoptotic process
cellular response to lithium ion
DNA biosynthetic process
establishment of integrated proviral latency
double-strand break repair via classical nonhomologous end joining
positive regulation of chromosome organization
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
protein binding
ATP binding
protein C-terminus binding
ligase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13009687.hg.1 ENST00000356922.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009687.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009687.hg.1 TC13000867.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164996.hg.1 TC1300009740.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 142541 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0