This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00934591_g1
See other KAT5 GE Assays ›
Gene Symbol:
KAT5
Gene Name:
lysine acetyltransferase 5
Gene Aliases:
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2
Chromosome Location:
Chr.11: 65711996 - 65719606 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00934591_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KAT5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206833.1 NP_001193762.1 3 - 4 649 114
NM_182709.2 NP_874368.1 4 - 5 550 114
GenBank mRNA AB209813.1 - 2 - 3 352 114
AK225100.1 - 4 - 5 499 114
AK226002.1 - 4 - 5 499 114
AK304664.1 - 3 - 4 399 114
BC000166.3 - 3 - 4 272 114
BC093032.1 - 4 - 5 317 30335942 114
BC143296.1 - 4 - 5 334 9051800 114
U67734.1 - 4 - 5 301 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100008034.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000640.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KAT5

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
DNA replication
double-strand break repair
double-strand break repair via nonhomologous end joining
transcription, DNA-templated
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
response to ionizing radiation
viral process
histone acetylation
androgen receptor signaling pathway
negative regulation of interleukin-2 production
regulation of growth
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cellular response to estradiol stimulus
regulation of signal transduction by p53 class mediator
positive regulation of protein acetylation
beta-catenin-TCF complex assembly
transcription coactivator activity
histone acetyltransferase activity
protein binding
acetyltransferase activity
metal ion binding
androgen receptor binding
repressing transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 ENST00000341318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 ENST00000352980.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 ENST00000377046.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 ENST00000525204.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 ENST00000530446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 ENST00000534650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 NM_001206833.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 NM_006388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 NM_182709.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 NM_182710.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11008020.hg.1 TC11000640.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000341318.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000352980.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000377046.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000525204.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000527544.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000528198.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000530446.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000530605.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000531880.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534104.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534293.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534650.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534681.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_001206833.1 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_006388.3 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_182709.2 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_182710.2 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000341318.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000352980.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000377046.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000525204.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000527544.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000528198.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000530446.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000530605.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000531880.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534104.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534293.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534650.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 ENST00000534681.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_001206833.1 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_006388.3 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_182709.2 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 NM_182710.2 Refseq
Affymetrix Human Clariom D Assay PSR1100158376.hg.1 TC1100008034.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0