This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00934711_g1
See other ATP5J2 GE Assays ›
Gene Symbol:
ATP5J2
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
Gene Aliases:
ATP5JL
Chromosome Location:
Chr.7: 99458161 - 99466201 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00934711_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5J2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003713.2 NP_001003713.1 2 - 3 177 85
NM_004889.3 NP_004880.1 2 - 3 195 85
GenBank mRNA AF047436.1 - 2 - 3 162 85
AF088918.1 - 2 - 3 148 85
AK223348.1 - 2 - 3 158 85
AY046911.1 - 2 - 3 137 85
BC003678.1 - 2 - 3 158 3826469 85
BU587946.1 - 2 - 3 165 85
CB155273.1 - 2 - 3 162 85
CD173604.1 - 2 - 3 199 85
CR456891.1 - 2 - 3 135 85
CR542155.1 - 2 - 3 135 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700013591.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07003366.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5J2

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
mitochondrial ATP synthesis coupled proton transport
ATPase activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012799.hg.1 ENST00000447543.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007641.hg.1 ENST00000451550.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 TC07003366.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 ENST00000292475.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 ENST00000449683.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 ENST00000544611.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07025095.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 TC07003366.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 ENST00000292475.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 ENST00000449683.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 ENST00000544611.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07025096.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009718.hg.1 ENST00000537967.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000359832.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000359832.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000359832.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000466753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000466753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000485011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000485011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000488775.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000488775.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000488775.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000523680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000524321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 ENST00000524321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000523680.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 ENST00000523680.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay PSR0700201266.hg.1 TC0700013591.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900007641.hg.1 ENST00000451550.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009718.hg.1 ENST00000537967.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay PSR0700201265.hg.1 TC0700013591.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000292475.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000359832.8 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000359832.8 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000359832.8 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000394186.3 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000414062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000449683.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000466753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000466753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000481899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000485011.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000485011.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000488775.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000488775.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000488775.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000491560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000523680.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000524321.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 ENST00000524321.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003713.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001003714.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_001039178.2 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay TC0700013591.hg.1 NM_004889.3 Refseq
Affymetrix Human Clariom D Assay TC0600012799.hg.1 ENST00000447543.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0