This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00935222_m1
See other PTPRK GE Assays ›
Gene Symbol:
PTPRK
Gene Name:
protein tyrosine phosphatase, receptor type K
Gene Aliases:
R-PTP-kappa
Chromosome Location:
Chr.6: 127968779 - 128520847 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00935222_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135648.2 NP_001129120.1 19 - 20 3180 111
NM_001291981.1 NP_001278910.1 21 - 22 3228 111
XM_006715537.2 XP_006715600.1 19 - 20 3228 111
XM_011536014.2 XP_011534316.1 21 - 22 3276 111
XM_011536015.2 XP_011534317.1 20 - 21 3267 111
XM_011536017.2 XP_011534319.1 22 - 23 3519 111
XM_011536018.2 XP_011534320.1 22 - 23 3192 111
XM_017011145.1 XP_016866634.1 20 - 21 3195 111
XM_017011146.1 XP_016866635.1 20 - 21 3144 111
XM_017011147.1 XP_016866636.1 20 - 21 3122 111
XM_017011149.1 XP_016866638.1 9 - 10 780 111
XM_017011150.1 XP_016866639.1 8 - 9 767 111
GenBank mRNA BC140775.1 - 19 - 20 2982 9021643 111
BC144512.1 - 19 - 20 2982 9053039 111
BC144513.1 - 21 - 22 3030 9053040 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein protein phosphatase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
cell adhesion
signal transduction
transforming growth factor beta receptor signaling pathway
negative regulation of cell proliferation
negative regulation of keratinocyte proliferation
cell migration
negative regulation of cell migration
neuron projection development
protein localization to cell surface
cellular response to reactive oxygen species
cellular response to UV
peptidyl-tyrosine dephosphorylation
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
focal adhesion assembly
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
beta-catenin binding
protein kinase binding
gamma-catenin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0