This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00936058_m1
See other PPP1R14A GE Assays ›
Gene Symbol:
PPP1R14A
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14A
Gene Aliases:
CPI-17, CPI17, PPP1INL
Chromosome Location:
Chr.19: 38251237 - 38256591 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00936058_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033256.2 NP_150281.1 3 - 4 568 95
GenBank mRNA AB056508.1 - 3 - 4 509 95
AF123763.1 - 3 - 4 359 95
AY050670.1 - 3 - 4 353 95
BC021089.1 - 3 - 4 333 5019808 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010595.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001496.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R14A

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein dephosphorylation
cellular response to drug
regulation of phosphorylation
negative regulation of catalytic activity
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020303.hg.1 ENST00000301242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020303.hg.1 ENST00000347262.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020303.hg.1 NM_001243947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020303.hg.1 NM_033256.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020303.hg.1 TC19001496.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 TC1900010595.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187106.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 TC1900010595.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187107.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 ENST00000591585.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay TC1900010595.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 NM_033256.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 TC1900010595.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000301242.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000347262.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000587515.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 ENST00000591291.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 NM_001243947.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187105.hg.1 NM_033256.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 176279 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0