| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023391.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023391.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023391.hg.1 |
TC04001686.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023392.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023392.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023392.hg.1 |
TC04001686.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400184108.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400184108.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400184108.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400184108.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400184108.hg.1 |
TC0400012252.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400184109.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400184109.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400184109.hg.1 |
ENST00000307720.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400184109.hg.1 |
NM_005038.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400184109.hg.1 |
TC0400012252.hg.1 |
Clariom D TC |