This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00936731_m1
See other NEXN GE Assays ›
Gene Symbol:
NEXN
Gene Name:
nexilin F-actin binding protein
Gene Aliases:
CMH20, NELIN
Chromosome Location:
Chr.1: 77888515 - 77946329 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00936731_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEXN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005271323.3 XP_005271380.1 5 - 6 630 110
XM_005271326.3 XP_005271383.1 4 - 5 465 110
GenBank mRNA AK300728.1 - 5 - 6 672 110
AK307487.1 - 4 - 5 465 110
BC055084.1 - 5 - 6 516 110
BC111395.1 - 2 - 3 170 4271572 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell migration
cardiac muscle fiber development
regulation of cytoskeleton organization
structural constituent of muscle
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0