Assay Details
| Gene Symbol: MAPRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014268.3 | NP_055083.1 | 1 - 2 | 299 | 72 | |
| GenBank mRNA | AB016823.1 | - | 1 - 2 | 234 | 72 | |
| AK315766.1 | - | 1 - 2 | 252 | 72 | ||
| BC007318.2 | - | 1 - 2 | 242 | 2989643 | 72 | |
| BT020086.1 | - | 1 - 2 | 119 | 72 | ||
| CR536545.1 | - | 1 - 2 | 119 | 72 | ||
| DC354326.1 | - | 1 - 2 | 299 | 72 | ||
| X94232.1 | - | 1 - 2 | 259 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB016823.1 transcripts.
Set Membership: |
|

MAPRE2
Genomic Map