This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00936761_m1
See other VRK1 GE Assays ›
Gene Symbol:
VRK1
Gene Name:
vaccinia related kinase 1
Gene Aliases:
PCH1, PCH1A
Chromosome Location:
Chr.14: 96797309 - 96881614 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00936761_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VRK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003384.2 NP_003375.1 2 - 3 272 72
XM_006720247.3 XP_006720310.1 2 - 3 310 72
XM_011537132.1 XP_011535434.1 1 - 2 171 72
XM_017021622.1 XP_016877111.1 1 - 2 171 72
XM_017021624.1 XP_016877113.1 2 - 3 310 72
XM_017021625.1 XP_016877114.1 2 - 3 351 72
XM_017021626.1 XP_016877115.1 2 - 3 310 72
GenBank mRNA AB000449.1 - 2 - 3 241 72
BC103761.1 - 2 - 3 250 5773877 72
BC112075.1 - 2 - 3 240 8327543 72
BC113510.1 - 2 - 3 240 8322562 72
BU535261.1 - 2 - 3 292 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400010649.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000608.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VRK1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
mitotic nuclear division
mitotic nuclear envelope disassembly
mitotic nuclear envelope reassembly
regulation of cell shape
histone H3-S10 phosphorylation
protein autophosphorylation
cell division
histone H3-T3 phosphorylation
Golgi disassembly
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
nucleosomal histone binding
histone kinase activity (H3-S10 specific)
histone kinase activity (H3-T3 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14007569.hg.1 ENST00000216639.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14007569.hg.1 NM_003384.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14007569.hg.1 TC14000608.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay PSR1400180516.hg.1 TC1400010649.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000216639.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000553683.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555067.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555351.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000555402.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 ENST00000557352.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq
Affymetrix Human Clariom D Assay TC1400010649.hg.1 NM_003384.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0