| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007578.hg.1 |
ENST00000216639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007578.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007578.hg.1 |
TC14000608.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007577.hg.1 |
ENST00000216639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007577.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007577.hg.1 |
TC14000608.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180534.hg.1 |
TC1400010649.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000553683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000555402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010649.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
ENST00000216639.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
ENST00000557352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
NM_003384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180532.hg.1 |
TC1400010649.hg.1 |
Clariom D TC |