This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00937856_m1
See other AGR3 GE Assays ›
Gene Symbol:
AGR3
Gene Name:
anterior gradient 3, protein disulphide isomerase family member
Gene Aliases:
AG-3, AG3, BCMP11, HAG3, PDIA18, hAG-3
Chromosome Location:
Chr.7: 16854711 - 16881989 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00937856_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176813.4 NP_789783.1 3 - 4 246 98
XM_005249627.2 XP_005249684.1 3 - 4 225 98
XM_011515152.2 XP_011513454.1 3 - 4 238 98
GenBank mRNA AY069977.1 - 2 - 3 211 98
AY359065.1 - 3 - 4 230 98
BC058284.1 - 3 - 4 251 4694757 98
BM984550.1 - 5 - 6 98
BU682778.1 - 5 - 6 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010358.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001164.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AGR3

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
dystroglycan binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07018224.hg.1 TC07001164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07018224.hg.1 ENST00000310398.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018224.hg.1 ENST00000402239.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018224.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay PSR0700173644.hg.1 TC0700010358.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000310398.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000402239.7 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000414935.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 ENST00000486448.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq
Affymetrix Human Clariom D Assay TC0700010358.hg.1 NM_176813.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0