This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00937984_m1
See other UNC119B GE Assays ›
Gene Symbol:
UNC119B
Gene Name:
unc-119 lipid binding chaperone B
Gene Aliases:
POC7B
Chromosome Location:
Chr.12: 120710435 - 120723640 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00937984_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC119B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080533.2 NP_001074002.1 4 - 5 682 93
GenBank mRNA AK126367.1 - 4 - 5 659 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200009140.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000950.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

protein transport
lipoprotein transport
cilium morphogenesis
protein binding
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12012334.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012334.hg.1 ENST00000537794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012334.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012334.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012334.hg.1 TC12000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12012336.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012336.hg.1 ENST00000537794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012336.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012336.hg.1 TC12000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12012337.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012337.hg.1 ENST00000537794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012337.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012337.hg.1 TC12000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000578311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000618898.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000618898.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 ENST00000618898.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009140.hg.1 NR_039849.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012338.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012338.hg.1 ENST00000537794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012338.hg.1 NM_001080533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012338.hg.1 TC12000950.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200167613.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200167613.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay PSR1200167613.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200167613.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay PSR1200167613.hg.1 TC1200009140.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200167612.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200167612.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay PSR1200167612.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200167612.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay PSR1200167612.hg.1 TC1200009140.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000344651.4 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000539658.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000578311.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000618898.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000618898.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 ENST00000618898.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NM_001080533.2 Refseq
Affymetrix Human Clariom D Assay TC1200009140.hg.1 NR_039849.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0